• USA Home
  • T6265 - Anti-Transferrin antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
T6265 Sigma

Anti-Transferrin antibody produced in goat

fractionated antiserum

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Immune Cell Signaling and Blood, Antibodies to Human Plasma Proteins,
species reactivity   human
application(s)   Ouchterlony double diffusion: 1:8
clone   polyclonal
antibody form   fractionated antiserum
storage temp.   2-8°C
Gene Information   human ... TF(7018)
biological source   goat
conjugate   unconjugated

Description

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Reconstitution

Reconstitute with 2 mL deionized water.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

T2027 Anti-Transferrin antibody produced in goat, whole antiserum

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?