EMAIL THIS PAGE TO A FRIEND
T6318 Sigma

Tyrphostin AG 34

≥98%, solid

Synonym: Tyrphostin A24

  • Empirical Formula (Hill Notation) C11H8N2O3

  • Molecular Weight 216.19

  •  MDL number MFCD01868809

  •  PubChem Substance ID 24278748

Purchase

Description

Biochem/physiol Actions

Inhibitor of tyrosine protein kinase in human colon cancer cell lines.

Caution

Photosensitive

Preparation Note

Tyrphostin AG 34 is soluble in DMSO and acetone.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?