• USA Home
  • T6566 - Tygon® 2275 high purity tubing
EMAIL THIS PAGE TO A FRIEND
T6566 Sigma
Tygon® 2275 high purity tubing
Purchase
Properties
Related Categories Labware, Manufacturer Browser, Norton, Tubing, Tygon 2275 tubing,
mfr. no.   Tygon
I.D. × O.D.   1/4 in. × 3/8 in.
Description
General description
Ideal for handling sensitive fluids such as pharmaceutical or biological solutions.
• Provides and uncompromising fluid path for sensitive solutions
• Low sorption maintains fluid integrity
• Plasticizer free, virtually no extractables
• Environmentally friendly, safe to dispose of through incineration
• Documented biocompatibility to the ISO 10993 Standard
• Meets USP Class VI, FDA, and NSF criteria
Legal Information
Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?