• USA Home
  • T6689 - Trypsin-EDTA solution

EMAIL THIS PAGE TO A FRIEND
T6689 Sigma

Trypsin-EDTA solution

0.25%

Purchase

Properties

Quality Level   GMP
concentration   0.25%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
YN5075000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?