EMAIL THIS PAGE TO A FRIEND
T6693 Sigma

Tyrphostin RG 14620

≥98%

Synonym: 2-(3′,5′-Dichlorophenyl)-1-(3″-pyridinyl)acrylonitrile

  • CAS Number 136831-49-7

  • Empirical Formula (Hill Notation) C14H8Cl2N2

  • Molecular Weight 275.13

  •  MDL number MFCD00236445

Purchase

Description

Biochem/physiol Actions

Non-phenolic tyrphostin-class tyrosine kinase inhibitor.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Price and Availability


DISCOVER Bioactive Small Molecules

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Phosphorylation is a ubiquitous cellular regulatory mechanism that mediates signal transduction pathways, which carry signals from the cell surface to the nucleus or cytoplasm, by altering the activi...
Keywords: Apoptosis, Bioactive small molecules, Cancer, Diseases, Inflammation, Metabolism, Neurodegenerative Diseases, Phosphorylations, Transduction

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?