• USA Home
  • T6695 - Tissue protein, cytoplasmic

EMAIL THIS PAGE TO A FRIEND
T6695 Sigma

Tissue protein, cytoplasmic

from human liver

Purchase

Description

Application

Highly purified cytoplasmic protein from whole tissue homogenates. Useful in electrophoresis, Western blotting, immunoassay, and other applications requiring source-specific, non-denatured protein.

Physical form

3 mg protein (approx. 10 mg protein/mL) in a solution containing 20 mM HEPES, pH 7.9, 1.5 mM MgCl2, 10 mM KCl, 1 mM EDTA, 200 mM sucrose, 10% glycerol, and a cocktail of protease inhibitor.

Price and Availability


Antibody Explorer

Explore with Confidence

Ultrafree Centrifugal Filter Units
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?