• USA Home
  • T6780 - Thermometer for developing tank
EMAIL THIS PAGE TO A FRIEND
T6780 Sigma
Thermometer for developing tank
Glass, floating
Purchase
Properties
Related Categories Labware, Temperature, Thermometers More...
availability   not available in EU
Description
Application
For monitoring temperature in developing tank without thermostatic mixing valve.
General description
Calibrated from 25-100 °F (-3-38 °C)
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?