• USA Home
  • T6825 - Director-Ready pT7-MAT-2 Set
EMAIL THIS PAGE TO A FRIEND
T6825 Sigma
Director-Ready pT7-MAT-2 Set
Purchase
Properties
storage temp.   −20°C
Description
General description
Director-Ready vectors are pre-digested plasmid vectors that are ready for ligation to compatible inserts generated using traditional methods or by using the Director Universal Cloning System PCR kit (Product No. RDC-1). Efficiencies for obtaining a positive recombinant clone are typically greater than 90% when used in conjunction with the Director Universal PCR System. The system also provides a simple, rapid and universal method to directionally clone and express PCR products.
Other Notes
Director-Ready pT7-MAT®-2 is a Hind III/Bgl II digest of the 4.8 bp Escherichia coli expression vector pT7-MAT-2 (Product No. E 5655). Director-Ready pT7-MAT-2 may be used to clone any insert containing a 5′ Hind III compatible end and a 3′ Bgl II compatible end.
Legal Information
Director is a trademark of Sigma-Aldrich Co. LLC
Director-Ready is a trademark of Sigma-Aldrich Co. LLC
MAT-Tag is a registered trademark of Sigma-Aldrich Co. LLC
pT7-MAT is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
Kit component only
Description
Product #
Add to Cart
Director-Ready pT7-MAT-2 SDS    
Verification Primer-T7F (with 5’ phosphorylation) SDS    
Verification Primer-T7R (with 5’ phosphorylation) SDS    
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?