• USA Home
  • T6944 - Anti-phospho-Tau (pThr212) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
T6944 Sigma

Anti-phospho-Tau (pThr212) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Alzheimer′s Disease, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology,
species reactivity   goat, rhesus monkey, mouse, baboon, bovine, human, rat
application(s)   western blot: 1:1,000
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous glycerol solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... MAPT(4137)
mouse ... Mapt(17762)
rat ... Mapt(29477)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

chemically synthesized phosphopeptide derived from the region of human tau that contains threonine212. The sequence is conserved in many species.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), 50% glycerol with 1 mg/mL bovine serum albumin (IgG, protease free), and 0.05% sodium azide.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?