• USA Home
  • T7002 - Thiocarbamyl Nitro Blue Tetrazolium

EMAIL THIS PAGE TO A FRIEND
T7002 Sigma

Thiocarbamyl Nitro Blue Tetrazolium

~90% (TLC)

Synonym: 2,2′-Di(p-nitrophenyl)-5,5′-di(p-thiocarbamylphenyl)-3,3′-(3,3′-dimethoxy-4,4′-biphenylene)ditetrazolium chloride, TC-NBT

Purchase

Properties

InChI Key   QIXARCLEZKFQSV-UHFFFAOYSA-O
assay   ~90% (TLC)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?