• USA Home
  • T7158 - Tetrabutylammonium bisulfate

EMAIL THIS PAGE TO A FRIEND
T7158 Sigma

Tetrabutylammonium bisulfate

~99%

Synonym: Tetrabutylammonium hydrogen sulfate

Purchase

Properties

InChI Key   SHFJWMWCIHQNCP-UHFFFAOYSA-M
assay   ~99%

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?