• USA Home
  • T7267 - 2,3,5-Triiodobenzoic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
T7267 Sigma

2,3,5-Triiodobenzoic acid sodium salt

  • CAS Number 17274-12-3

  • Linear Formula C7H2I3O2Na

  • Molecular Weight 521.79

  •  PubChem Substance ID 329826863

Purchase

Properties

InChI Key   CXMSAYPXSMPSBH-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
DH9360000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

T5910 2,3,5-Triiodobenzoic acid, BioReagent, plant cell culture tested, ≥97% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?