• USA Home
  • T7283 - Tetrabutylammonium perchlorate

EMAIL THIS PAGE TO A FRIEND
T7283 Sigma

Tetrabutylammonium perchlorate

  • CAS Number 1923-70-2

  • Linear Formula (CH3CH2CH2CH2)4N(ClO4)

  • Molecular Weight 341.91

  •  Beilstein Registry Number 3579274

  •  EC Number 217-655-5

  •  MDL number MFCD00038722

  •  PubChem Substance ID 24900443

Purchase

Properties

Related Categories Biochemicals and Reagents, Chemical Synthesis, Detergents, Detergents N to Z, Oxidizing Agents,
InChI Key   KBLZDCFTQSIIOH-UHFFFAOYSA-M

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1479 5.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

86885 Tetrabutylammonium perchlorate, ≥98.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?