• USA Home
  • T7306 - Trp-Gly hydrochloride

EMAIL THIS PAGE TO A FRIEND
T7306 Sigma

Trp-Gly hydrochloride

  • CAS Number 63193-13-5

  • Empirical Formula (Hill Notation) C13H15N3O3 · HCl

  • Molecular Weight 297.74

  •  MDL number MFCD00678206

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
storage temp.   −20°C

Description

Other Notes

Dipeptide found in human pituitary gland.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?