• USA Home
  • T7516 - Anti-human polyvalent immunoglobulins TRITC CONJUGATE

EMAIL THIS PAGE TO A FRIEND
T7516 Sigma

Anti-human polyvalent immunoglobulins TRITC CONJUGATE

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, HU-HZ, Primary Antibodies
clone   polyclonal
conjugate   TRITC conjugate
storage temp.   −20°C

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?