EMAIL THIS PAGE TO A FRIEND
T7821 Sigma

[Ser409]-Tau

>85% (HPLC)

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
assay   >85% (HPLC)
mol wt   mol wt 1660
solubility   H2O: soluble
storage temp.   −20°C

Description

Application

Negative control peptide for the [pSer409]-Tau rabbit polyclonal antibody.

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?