• USA Home
  • T7890 - Tetrahydrodeoxyaflatoxin B1

EMAIL THIS PAGE TO A FRIEND
T7890 Sigma

Tetrahydrodeoxyaflatoxin B1

~97%

Purchase

Properties

assay   ~97%
storage temp.   −20°C

Description

Preparation Note

Tetrahydrodeoxyaflatoxin B1 has been used for enzyme immunoassays (EIA) in human urine samples.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?