• USA Home
  • T7895 - 2′,3′,5′-Tri-O-acetyladenosine

EMAIL THIS PAGE TO A FRIEND
T7895 Sigma

2′,3′,5′-Tri-O-acetyladenosine

≥98%

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   GCVZNVTXNUTBFB-XNIJJKJLSA-N
assay   ≥98%
storage temp.   −20°C

Description

Packaging

500 mg in poly bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?