• USA Home
  • T7937 - Anti-Tissue Inhibitor of Metalloproteinase-2, C-Terminal antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
T7937 Sigma

Anti-Tissue Inhibitor of Metalloproteinase-2, C-Terminal antibody produced in rabbit

~1 mg/mL, buffered aqueous solution, affinity isolated antibody

Synonym: Anti-TIMP-2

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies against Adhesion, Attachment, and Matrix Factors, Antibodies for Cell Biology, Antibodies for Hematopoietic Stem Cells,
species reactivity   rabbit, rat, mouse, human, guinea pig, bovine
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): 10-20 μg/mL using pretreated tissue sections
  western blot: 5-10 μg/mL using concentrated cell culture medium from a stimulated human cell line (M2928) or human fetal lung cells
clone   polyclonal
concentration   ~1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt 21 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... TIMP2(7077)
mouse ... Timp2(21858)
rat ... Timp2(29543)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to the C-terminus of human tissue inhibitor of metalloproteinase-2.

Physical form

Solution in 10 mM phosphate buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.09% sodium azide.

Specificity

By immunoblotting against the reduced protein, the antibody reacts with a band at approximately 21 kDa. It does not perform well with non-reduced TIMP-2.

Price and Availability


Antibody Explorer

Explore with Confidence

Biomedical Applications
Safety & Documentation

Safety Information

Symbol  GHS08  GHS08
Signal word  Danger
Hazard statements  H319-H334
Precautionary statements  P261-P305 + P351 + P338-P342 + P311
Personal Protective Equipment  Eyeshields, Faceshields, full-face respirator (US), Gloves, multi-purpose combination respirator cartridge (US), type ABEK (EN14387) respirator filter
RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?