• USA Home
  • T7942 - Standard PCR microtubes
EMAIL THIS PAGE TO A FRIEND
T7942 Sigma
Standard PCR microtubes
PCR tube (Domed cap), volume 0.65 mL
Purchase
Properties
Related Categories Labware, Molecular Biology Supplies, PCR (Polymerase Chain Reaction) Supplies, PCR Tubes, Standard PCR tubes More...
material   cap domed
  natural
  polypropylene
  tube V-bottom
feature   RNase and DNase free
packaging   pack of 1,000 ea
volume   0.65 mL
Description
General description
Precision moulded from virgin polypropylene and clean room produced.
Certified free from DNase, RNase and human genomic DNA contamination.
Suitable for 0.5 mL thermal cycler blocks
Integral snap-shut domed cap
Price and Availability
Your Key to Success
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?