• USA Home
  • T8322 - Anti-Tissue Inhibitor of Metalloproteinase-1, First Loop antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
T8322 Sigma

Anti-Tissue Inhibitor of Metalloproteinase-1, First Loop antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym: Anti-TIMP-1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies against Adhesion, Attachment, and Matrix Factors, Antibodies for Cell Biology, Antibodies for Hematopoietic Stem Cells,
species reactivity   human
application(s)   immunohistochemistry (frozen sections): suitable
  immunoprecipitation: suitable
  indirect ELISA: suitable
  western blot (chemiluminescent): 1:5,000
  western blot: 1:1,000
clone   polyclonal
concentration   ~1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous glycerol solution
mol wt   antigen mol wt 29 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... TIMP1(7076)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to the first loop of human TIMP-1

Physical form

Solution in 0.01 M phosphate buffered saline, containing 50% glycerol and 0.05% sodium azide.

Specificity

Reacts with reduced and non-reduced TIMP-1

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Price and Availability


Antibody Explorer

Explore with Confidence

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany  1
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?