• USA Home
  • T8380 - 2,3,5-Tri-O-benzyl-β-L-arabinofuranose

EMAIL THIS PAGE TO A FRIEND
T8380 Sigma

2,3,5-Tri-O-benzyl-β-L-arabinofuranose

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates P-Z, Monosaccharide More...
InChI Key   NAQUAXSCBJPECG-SSUZURRFSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?