EMAIL THIS PAGE TO A FRIEND
T8407 Sigma

TIP39

  • Empirical Formula (Hill Notation) C202H325N61O54S

  • Molecular Weight 4504.18

  •  MDL number MFCD02689182

  •  PubChem Substance ID 329826880

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Parathyroid Hormone Fragments, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   KKPNKVPBWQJZOL-UHFFFAOYSA-N
storage temp.   −20°C
Gene Information   human ... PTH2(113091)

Description

Amino Acid Sequence

Ser-Leu-Ala-Leu-Ala-Asp-Asp-Ala-Ala-Phe-Arg-Glu-Arg-Ala-Arg-Leu-Leu-Ala-Ala-Leu-Glu-Arg-Arg-His-Trp-Leu-Asn-Ser-Tyr-Met-His-Lys-Leu-Leu-Val-Leu-Asp-Ala-Pro

Biochem/physiol Actions

Parathyroid hormone 2 receptor ligand

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?