• USA Home
  • T8412 - Barnant hand-held thermometer
EMAIL THIS PAGE TO A FRIEND
T8412 Sigma
Barnant hand-held thermometer
Model 120 EXI
Purchase
Properties
mfr. no.   Barnant
measuring range   temperature, accuracy: 0.4 °C, (throughout range)
battery power   9 V
Description
General description
All-solid state, microprocessor controlled meters accept a wide variety of thermocouple junction types and offer unmatched flexibility, range and accuracy.
• 4-digit LCD with selectable resolution (0.1° or 1°), reading °C or °F (auto-select above 1000°)
• Accepts ANSI standard thermocouple probes with mini-connectors (see Probes for specific ranges)
• Calibrated against NBS curves for each of the thermocouple types
• Built-in tilt stand for desktop use
Linkage
Same specifications as Model 115, but with an "intrinsically safe" design for hazardous locations (conforms to CSA Certification Class 1, Division 1, Groups A, B, C, and D).
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?