• USA Home
  • T8428 - Trypsin Inhibitor−Celite®

EMAIL THIS PAGE TO A FRIEND
T8428 Sigma

Trypsin Inhibitor−Celite®

powder, average diameter 1.8 μm, trypsin inhibitor from Phaseolus limensis (lima bean)

Purchase

Properties

Related Categories Affinity Chromatography, Molecular Biology, Protein Chromatography, Proteomics, Specialty Resins More...
biological source   trypsin inhibitor from Phaseolus limensis (lima bean)
form   powder
extent of labeling   50-75 mg per g solid
matrix   powdered Celite
matrix spacer   8 atoms
pore size   1.8 μm average diameter
capacity   20-80 mg/mg, solid binding capacity (trypsin)(with an activity of 10,000 BAEE units per mg)
storage temp.   2-8°C

Description

Application

Trypsin inhibitor Celite is used for protein chromatography, affinity chromatography and specialty resins. Trypsin inhibitor Celite has been used to determine that corn trypsin inhibitor decreases tissue-type plasminogen activator-mediated fibrinolysis of human plasma. Trypsin inhibitor celite has also been used in numerous studies assessing activtated coagulation time.

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?