• USA Home
  • T8529 - Trizma® hydrochloride

EMAIL THIS PAGE TO A FRIEND
T8529 Sigma

Trizma® hydrochloride

for electrophoresis, ≥99%

Synonym: TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride

  • CAS Number 1185-53-1

  • Linear Formula NH2C(CH2OH)3 · HCl

  • Molecular Weight 157.60

  •  Beilstein Registry Number 3675235

  •  EC Number 214-684-5

  •  MDL number MFCD00012590

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

grade   for electrophoresis
InChI Key   QKNYBSVHEMOAJP-UHFFFAOYSA-N
assay   ≥99%
useful pH range   7.0 - 9.0
pKa (25 °C)   8.1
cation traces   heavy metals (as Pb): ≤5 ppm
absorption   A40%/290 ≤0.05

Description

Application

The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution.
For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

Trizma is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

T5941 Trizma® hydrochloride, BioPerformance Certified, cell culture tested, ≥99.0% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?