• USA Home
  • T8655 - Trizma® phosphate monobasic

EMAIL THIS PAGE TO A FRIEND
T8655 Sigma

Trizma® phosphate monobasic

BioReagent, for molecular biology, ≥98%

Synonym: Mono(tris[hydroxymethyl]aminomethane) phosphate, Mono-Tris phosphate, Tris phosphate, Tris(hydroxymethyl)aminomethane phosphate

Purchase

Properties

grade   for molecular biology
InChI Key   JLEXUIVKURIPFI-UHFFFAOYSA-N
product line   BioReagent
assay   ≥98%
cation traces   heavy metals (as Pb): <5 ppm
foreign activity   DNase, none detected
  Protease, none detected
  RNase, none detected

Description

Legal Information

Trizma is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

93348 Trizma® phosphate monobasic, BioXtra, ≥99.0% (NT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?