• USA Home
  • T8661 - Transforming Growth Factor-α Fragment 34-43 Rat

EMAIL THIS PAGE TO A FRIEND
T8661 Sigma

Transforming Growth Factor-α Fragment 34-43 Rat

≥97% (HPLC)

  • CAS Number 97474-88-9

  • Empirical Formula (Hill Notation) C44H67N15O13S2

  • Molecular Weight 1078.23

  •  MDL number MFCD00151765

  •  PubChem Substance ID 24900551

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   MIHUXUMZEJHQNC-UHFFFAOYSA-N
assay   ≥97% (HPLC)
composition   Peptide content, ~65%
storage temp.   −20°C
Gene Information   rat ... Tgfa(24827)

Description

Amino Acid Sequence

Cys-His-Ser-Gly-Tyr-Val-Gly-Val-Arg-Cys [Disulfide bridge: 1–10]

Packaging

250 μg in serum bottle

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?