EMAIL THIS PAGE TO A FRIEND
T8677 Sigma

TTA-386

≥97% (HPLC)

Synonym: Hexamethyleneiminocarbonyl-Leu-D-Trp-D-Ala-β-Ala-Tyr-D-Phe

Purchase

Properties

InChI Key   QVTBFVFPWXEIOX-VYAQOJDXSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

An endothelin ETA receptor-selective antagonist to ET-1.

Packaging

Package size based on peptide content

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?