• USA Home
  • T8800 - Trichrome TISSUE-TROL

EMAIL THIS PAGE TO A FRIEND
T8800 Sigma

Trichrome TISSUE-TROL

from human liver

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, TISSUE-TROL Control Slides More...
Quality Level   GMP-IVD
biological source   from human liver
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use

Description

Application

Suitable for use with Sigma Trichrome Stains, Procedure Nos. HT10 and HT15.

Legal Information

TISSUE-TROL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

TTR012 Trichrome TISSUE-TROL Control Slides, from human liver

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?