• USA Home
  • T8940 - cDNA Probe α-Tubulin from rat

EMAIL THIS PAGE TO A FRIEND
T8940 Sigma

cDNA Probe α-Tubulin from rat

Purchase

Description

Application

Genes that are highly conserved and expressed in a wide variety of tissue types are useful as reference "house-keeping" genes. They allow determination of relative expression levels of other genes of interest.

General description

GenBank® Accession Number: V01227.

Other Notes

A rat α-tubulin cDNA probe is a 0.48-kb fragment of rat α-tubulin (bp 975-1421).

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

required but not provided

Product #

Description

Add to Cart

W4502 Water, Molecular Biology Reagent

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?