• USA Home
  • T9072 - Tissue Inhibitor of Metalloproteinase-2 from human fibroblasts

EMAIL THIS PAGE TO A FRIEND
T9072 Sigma

Tissue Inhibitor of Metalloproteinase-2 from human fibroblasts

>95% (SDS-PAGE), buffered aqueous glycerol solution

Synonym: TIMP-2

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes, Cell Signaling and Neuroscience,
assay   >95% (SDS-PAGE)
form   buffered aqueous glycerol solution
mol wt   mol wt ~23 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... TIMP2(7077)

Description

Biochem/physiol Actions

TIMP-2 has a greater binding efficiency to MMP-2 than the other MMPs. Although TIMP-2 is an inhibitor of MMP-2, it is also required at low concentrations for the activation of MMP-2.

Physical form

Supplied in 0.01 M phosphate buffered saline and 150 mM sodium chloride containing 50% glycerol.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

T1077 Tissue Inhibitor of Metalloproteinase-2 human, recombinant, expressed in CHO cells, >95% (SDS-PAGE), lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?