• USA Home
  • T9150 - Testosterone-3-(O-carboxymethyl)oxime–N-(4-aminobutyl)-N-ethylisoluminol conjugate

EMAIL THIS PAGE TO A FRIEND
T9150 Sigma

Testosterone-3-(O-carboxymethyl)oxime–N-(4-aminobutyl)-N-ethylisoluminol conjugate

Synonym: Testosterone-3-CMO: ABEI

  • CAS Number 82546-60-9

  • Empirical Formula (Hill Notation) C35H49N5O5

  • Molecular Weight 619.79

  •  PubChem Substance ID 329826901

Purchase

Properties

InChI Key   JSEWGWPMIFTGAC-WOGQHTPQSA-N
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
storage temp.   −20°C

Description

Application

Used in chemiluminescent immunoassays.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?