• USA Home
  • T9306 - Tag-72 antigen from human fluids

EMAIL THIS PAGE TO A FRIEND
T9306 Sigma

Tag-72 antigen from human fluids

buffered aqueous solution

Synonym: Cancer-associated Ca-72-4

Purchase

Properties

Related Categories Cancer Markers and Antibodies, Cancer Research, Cell Biology, Cell Signaling and Neuroscience, Immune Cell Signaling and Blood,
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C

Description

Quality

Partially purified

Unit Definition

One unit is an arbitrary amount related to a reference antigen preparation using CIS-RIA method.

Physical form

Solution in 0.15 M phosphate buffered saline, pH 7.4, containing 0.1% sodium azide.

Biochem/physiol Actions

Expression of TAG-72 was shown to be induced by interferon α treatment in patients with colorectal cancer.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?