• USA Home
  • T9400 - Triosephosphate Isomerase from rabbit muscle

EMAIL THIS PAGE TO A FRIEND
T9400 Sigma

Triosephosphate Isomerase from rabbit muscle

Prestained

Purchase

Properties

Related Categories Cell Biology, General Metabolic Enzymes, General Metabolic Enzymes I-Z, Metabolomics
mol wt   unstained protein mol wt 26,600 Da
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?