• USA Home
  • T9446 - Nunc® sealing tape for multiwell plates
EMAIL THIS PAGE TO A FRIEND
T9446 Sigma
Nunc® sealing tape for multiwell plates
white polyester
Purchase
Properties
Related Categories Labware, Manufacturer Browser, Multiwell Plates, Nunc, Nunc Multiwell Plate Seals,
material   white polyester
description   acrylate adhesive
sterility   sterile
mfr. no.   Nunc 235305
Description
General description
A complete line of sealing tapes for use with MicroWell plates in a wide range of applications. The performance of these tapes has been characterized under a variety of conditions, including an extensive study of the evaporation of well contents and the adherence of tapes to plates manufactured in various polymers.
Featuring 3M adhesive technology, sealing tapes provide a highly effective seal, preventing evaporation and eliminating contamination.
Specific tapes are intended for use in General Assays, Genomics, Compound and Library Storage, Bio-Analytical Assays, High Throughput Screening and Drug Discovery applications.
• Provide an effective seal on polypropylene, polystyrene, and polycarbonate microwell plates
• Compatible with aqueous solutions and organic solvents
• Tested for use between -80 and +110°C
• Pre-cut, plate-sized sheets
• Protective backing
• Minimal evaporation of plate contents
• Eliminates well-to-well cross contamination
• Easy to handle delivery system
• Provided with removable tabs
• For storage, PCR, microscopy, culture, protection
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?