• USA Home
  • T9663 - Tygon® formula R-3603 laboratory tubing
EMAIL THIS PAGE TO A FRIEND
T9663 Sigma
Tygon® formula R-3603 laboratory tubing
I.D. × O.D. 5/8 in. × 7/8 in.
Purchase
Properties
Related Categories Labware, Manufacturer Browser, Norton, Tubing, Tygon Tubing,
mfr. no.   Tygon
I.D. × O.D.   5/8 in. × 7/8 in.
wall size   1/8 in.
Description
General description
Clear, general-purpose laboratory tubing. Ideal for peristaltic pumps, autoclavable to 248°F for 30 min. Temp. range: -49 to 158°F. Heavy walled Tygon is suitable for vacuum use.
• Resists most inorganic chemicals
• Resistant to cracking and aging
• Low vapor pressure
• Lasts longer than rubber tubing
Legal Information
Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?