• USA Home
  • T9750 - Thymidine 5′-monophosphate α-naphthyl ester ammonium salt

EMAIL THIS PAGE TO A FRIEND
T9750 Sigma

Thymidine 5′-monophosphate α-naphthyl ester ammonium salt

Synonym: α-Naphthyl 5′-TMP

Purchase

Properties

impurities   α-naphthol and α-naphthyl phosphate., essentially free
storage temp.   −20°C

Description

Substrates

A chromogenic and histochemical substrate for phosphodiesterase from kidney and snake venom (yields thymidine 5′-phosphate and α-naphthol). Resists hydrolysis by spleen phosphodiesterase, DNase I, DNase II, and RNase.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?