• USA Home
  • TMS17P - Trimethylsilyl (tms) sugar derivatives

EMAIL THIS PAGE TO A FRIEND
TMS17P Sigma

Trimethylsilyl (tms) sugar derivatives

~98% (GLC)

Purchase

Properties

assay   ~98% (GLC)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?