• USA Home
  • TOX3 - In Vitro Toxicology Assay Kit, Acid Phosphatase based

EMAIL THIS PAGE TO A FRIEND
TOX3 Sigma

In Vitro Toxicology Assay Kit, Acid Phosphatase based

Purchase

Properties

usage   1 kit sufficient for 1,000 tests
storage temp.   −20°C
Gene Information   human ... ACP1(52)

Description

Application

For spectrophotometric measurement of biomass (viable and nonviable cells) by membrane associated acid phosphatase activity. Absorbance of converted dye is measured at a wavelength of 405 nm.

Biochem/physiol Actions

Conversion of p-nitrophenyl phosphate to a colored compound by acid phosphatase.

Price and Availability

Standard component

Description

Product #

Add to Cart

acid phosophatase substrate 5 x 60 mg SDS    
assay buffer 25 mL SDS    
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?