EMAIL THIS PAGE TO A FRIEND
U0258 Sigma

Ubenimex

solid

Synonym: Bestatin hydrochloride, NK-421, [S-(R*,S*)]-N-(3-Amino-2-hydroxy-1-oxo-4-phenylbutyl)-L-leucine

  • CAS Number 58970-76-6

  • Empirical Formula (Hill Notation) C16H24N2O4

  • Molecular Weight 308.37

  •  EC Number 261-529-2

  •  PubChem Substance ID 329827590

Purchase

Properties

InChI Key   VGGGPCQERPFHOB-RDBSUJKOSA-N
form   solid
mol wt   apparent mol wt 308.38

Description

Application

Antitumor and antibiotic

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
OH2915000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?