• USA Home
  • U0757 - Anti-Urotensin II antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
U0757 Sigma

Anti-Urotensin II antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Neurotransmission, Antibodies to Neuropeptides, Neuroscience,
species reactivity   human
application(s)   indirect ELISA: suitable
  microarray: suitable
  radioimmunoassay: 1:200,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~1.4 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... UTS2(10911)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic human Urotensin-II peptide conjugated to KLH.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?