• USA Home
  • U1632 - Ubiquitin, Methylated from bovine erythrocytes

EMAIL THIS PAGE TO A FRIEND
U1632 Sigma

Ubiquitin, Methylated from bovine erythrocytes

semisynthetic, ≥90% (SDS-PAGE), essentially salt-free, lyophilized powder

Purchase

Description

Preparation Note

Methylated ubiquitin is soluble in water (at least 0.3 mg/ml).

Prepared from ubiquitin by reductive methylation of primary amino groups.

Application

Methylated ubiquitin can be ligated to proteins but cannot form polyubiquitin chains. It inhibits the elongation of polyubiquitin chains and the degradation of ubiquitin-dependent protein.

Biochem/physiol Actions

Methylated ubiquitin is known to block the degradation of cyclin in extracts of clam embryos.

Price and Availability


Better Together

Discover your Target

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?