EMAIL THIS PAGE TO A FRIEND
U1755 Sigma

Urapidil

Purchase

Properties

InChI Key   ICMGLRUYEQNHPF-UHFFFAOYSA-N

Description

Biochem/physiol Actions

α-Adrenergic antagonist; peripheral vasodilator;

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
YQ9862000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

U100 Urapidil hydrochloride, solid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?