EMAIL THIS PAGE TO A FRIEND
U2504 Sigma

Uramil

~95%

Synonym: 5-Amino-2,4,6-trihydroxypyrimidine, 5-Aminobarbituric acid

  • CAS Number 118-78-5

  • Empirical Formula (Hill Notation) C4H5N3O3

  • Molecular Weight 143.10

  •  EC Number 204-277-0

Purchase

Properties

assay   ~95%
color   off-white
solubility   1 M NaOH: soluble, clear

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?