• USA Home
  • U2627 - Uridine 5′-diphosphohexanolamine sodium salt

EMAIL THIS PAGE TO A FRIEND
U2627 Sigma

Uridine 5′-diphosphohexanolamine sodium salt

~95%

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleotides, Oligonucleotides, Nucleotide Analogs More...
InChI Key   YMFHJYFCJHCAOT-UHFFFAOYSA-N
assay   ~95%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?