• USA Home
  • U3250 - Uricase from Porcine Liver

EMAIL THIS PAGE TO A FRIEND
U3250 Sigma

Uricase from Porcine Liver

~2 units/mL, ammonium sulfate suspension, 4,000-8,000 units/g protein (biuret)

Purchase

Properties

form   ammonium sulfate suspension
concentration   ~2 units/mL
storage temp.   2-8°C

Description

Unit Definition

One unit will convert 1.0 μmole of uric acid to allantoin per min at pH 8.5 at 25°C, unless otherwise indicated.

Physical form

Suspension in 2.0 M (NH4)2SO4

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YU7100000

Documents

Certificate of Analysis

Protocols & Articles

Articles

Ammonium Sulfate Suspensions - Technical Note

Note that for an ammonium sulfate suspension, most of the enzyme will be in solid form. It is likely that only negligible amounts of enzyme will be in the ammonium sulfate solution.
Keywords: Enzyme activity

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?