• USA Home
  • U3500 - Uricase from bovine kidney

EMAIL THIS PAGE TO A FRIEND
U3500 Sigma

Uricase from bovine kidney

~15 units/g protein (biuret)

Purchase

Properties

storage temp.   −20°C

Description

Unit Definition

One unit will convert 1.0 μmole of uric acid to allantoin per min at pH 8.5 at 25°C, unless otherwise indicated.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YU7100000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?