• USA Home
  • U3760 - Universal Immunoassay Buffer 10X

EMAIL THIS PAGE TO A FRIEND
U3760 Sigma

Universal Immunoassay Buffer 10X

Purchase

Description

Application

Wash 3-5 times with this buffer in between steps for immunoassay protocols.

General description

Immunoassay buffer is supplied as 10× solution. It is used as 1× in all immunoassays as a washing buffer and can be used as dilution buffer for antibodies; however it does not contain any carrier proteins. This buffer does not contain any phosphate, sodium azide or mercury preservatives and can be used for dilution of all antibodies, including peroxidase conjugates and antibodies to phosphoproteins. Not suitable for dilution of antibodies to S100 proteins.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?