EMAIL THIS PAGE TO A FRIEND
U4128 Sigma

Urea

BioReagent, suitable for cell culture

Synonym: Carbamide, Carbonyldiamide

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Culture, Chaotropic Agents, Denaturation, Miscellaneous Reagents and Supplements,
InChI Key   XSQUKJJJFZCRTK-UHFFFAOYSA-N
product line   BioReagent
form   pellets
mp   132-135 °C(lit.)
solubility   H2O: soluble100 mg/mL
density   1.335 g/mL at 25 °C(lit.)
suitability   suitable for cell culture
Gene Information   human ... CA1(759), CA2(760)
rat ... Ppm1a(24666)

Description

Application

Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Price and Availability


Filter out uncertainty.

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
YR6250000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?